COMH4211C445 Combi-Blocks, Inc.

3-Fluoro-4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-benzoic acid tert-butyl ester

  • MDL number MFCD30486229
  • Cas Number : 1980783-92-3
  • Molecular Weight : 322.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9224

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9224
Availability details for
Quantity
  • Availability
  • Add to Cart