COMH4211C449 Combi-Blocks, Inc.

tert-Butyl 4-(2-(4-aminophenoxy)ethyl)piperazine-1-carboxylate

  • MDL number MFCD11848965
  • Cas Number : 862874-77-9
  • Molecular Weight : 321.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9231

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9231
Availability details for
Quantity
  • Availability
  • Add to Cart