COMH4211C44D Combi-Blocks, Inc.

4-(3-Fluoro-propoxy)-2-methyl-benzaldehyde

  • MDL number MFCD31628775
  • Cas Number : 1562412-74-1
  • Molecular Weight : 196.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9237

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9237
Availability details for
Quantity
  • Availability
  • Add to Cart