COMH4211C44F Combi-Blocks, Inc.

1,1-Dimethylethyl (2-bromophenyl)sulfonyl(cyclopropyl)carbamate

  • MDL number MFCD31613920
  • Cas Number : 503069-04-3
  • Molecular Weight : 376.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9243

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9243
Availability details for
Quantity
  • Availability
  • Add to Cart