COMH4211C451 Combi-Blocks, Inc.

N-Benzyl-5-bromo-2-methoxybenzamide

  • MDL number MFCD06091277
  • Cas Number : 831252-42-7
  • Molecular Weight : 320.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9246

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9246
Availability details for
Quantity
  • Availability
  • Add to Cart