COMH4211C457 Combi-Blocks, Inc.

N-Boc-o-allyl-n-ethylhydroxylamine

  • MDL number MFCD31628733
  • Molecular Weight : 201.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9264

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9264
Availability details for
Quantity
  • Availability
  • Add to Cart