COMH4211C45B Combi-Blocks, Inc.

2-Benzoyloxyethanethioamide

  • MDL number MFCD07783818
  • Cas Number : 40516-65-2
  • Molecular Weight : 195.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9275

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9275
Availability details for
Quantity
  • Availability
  • Add to Cart