COMH4211C460 Combi-Blocks, Inc.

(5-Chloro-2-methoxybenzyl)(triphenyl)phosphonium bromide

  • MDL number MFCD31561554
  • Cas Number : 1071618-35-3
  • Molecular Weight : 497.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9292

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9292
Availability details for
Quantity
  • Availability
  • Add to Cart