COMH4211C463 Combi-Blocks, Inc.

N,N-Dimethyl-4-ethenylbenzamide

  • MDL number MFCD28363968
  • Cas Number : 2681-30-3
  • Molecular Weight : 175.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9299

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9299
Availability details for
Quantity
  • Availability
  • Add to Cart