COMH4211C464 Combi-Blocks, Inc.

(3-Fluoro-4-propoxyphenyl)methanamine

  • MDL number MFCD11619340
  • Cas Number : 1094510-30-1
  • Molecular Weight : 183.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9300

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9300
Availability details for
Quantity
  • Availability
  • Add to Cart