COMH4211C483 Combi-Blocks, Inc.

[4-(2-Methoxy-ethoxy)-pyrazol-1-yl]-acetic acid

  • MDL number MFCD31579322
  • Cas Number : 1890862-88-0
  • Molecular Weight : 200.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9313

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9313
Availability details for
Quantity
  • Availability
  • Add to Cart