COMH4211C484 Combi-Blocks, Inc.

1-[4-(2-tert-Butylphenyl)piperazin-1-yl]carbonylcyclopropanecarboxylic acid

  • MDL number MFCD31613745
  • Cas Number : 1252656-29-3
  • Molecular Weight : 330.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9316

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9316
Availability details for
Quantity
  • Availability
  • Add to Cart