COMH4211C488 Combi-Blocks, Inc.

3-Methoxymethoxy-2,2-dimethylpropionic acid

  • MDL number MFCD24502153
  • Cas Number : 1350551-92-6
  • Molecular Weight : 162.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9333

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9333
Availability details for
Quantity
  • Availability
  • Add to Cart