COMH4211D184 Combi-Blocks, Inc.

3-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-4-(2,2,2-trifluoro-ethoxy)-benzonitrile

  • MDL number MFCD31381698
  • Molecular Weight : 327.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9344

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9344
Availability details for
Quantity
  • Availability
  • Add to Cart