COMH4211D186 Combi-Blocks, Inc.

Piperidine-1-carboxylic acid (3-bromo-phenyl)-amide

  • MDL number MFCD22205604
  • Cas Number : 60465-16-9
  • Molecular Weight : 283.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9347

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9347
Availability details for
Quantity
  • Availability
  • Add to Cart