COMH4211D188 Combi-Blocks, Inc.

3-(4-Amino-2-methyl-phenyl)-propionic acid ethyl ester

  • MDL number MFCD28813013
  • Cas Number : 865139-47-5
  • Molecular Weight : 207.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9354

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9354
Availability details for
Quantity
  • Availability
  • Add to Cart