COMH4211D18A Combi-Blocks, Inc.

Benzeneacetamide, 2-methyl-n-[4-(5-oxazolyl)phenyl]-

  • MDL number MFCD30486222
  • Cas Number : 1626923-39-4
  • Molecular Weight : 292.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9357

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9357
Availability details for
Quantity
  • Availability
  • Add to Cart