COMH4211D18E Combi-Blocks, Inc.

[1-(2-Amino-6-chloro-phenyl)-ethyl]-carbamic acid tert-butyl ester

  • MDL number MFCD31579547
  • Cas Number : 2270908-83-1
  • Molecular Weight : 270.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9367

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9367
Availability details for
Quantity
  • Availability
  • Add to Cart