COMH4211D192 Combi-Blocks, Inc.

5-Bromo-2-(2,2,2-trifluoroethoxy)benzoic acid

  • MDL number MFCD11194954
  • Cas Number : 847786-18-9
  • Molecular Weight : 299.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9377

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9377
Availability details for
Quantity
  • Availability
  • Add to Cart