COMH4211D19B Combi-Blocks, Inc.

tert-Butyl 2-(3-iodo-1h-pyrazol-1-yl)-2-methylpropanoate

  • MDL number MFCD31579311
  • Cas Number : 1650548-57-4
  • Molecular Weight : 336.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9399

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9399
Availability details for
Quantity
  • Availability
  • Add to Cart