COMH4211D19E Combi-Blocks, Inc.

2-(3-(4-Bromophenyl)propyl)isoindoline-1,3-dione

  • MDL number MFCD31561555
  • Cas Number : 1054479-09-2
  • Molecular Weight : 344.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9403

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9403
Availability details for
Quantity
  • Availability
  • Add to Cart