COMH4211D1A3 Combi-Blocks, Inc.

1-(1-(2-Chlorophenyl)propyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1h-pyrazole

  • MDL number MFCD31583077
  • Cas Number : 1604037-07-1
  • Molecular Weight : 346.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9412

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9412
Availability details for
Quantity
  • Availability
  • Add to Cart