COMH4211D1A4 Combi-Blocks, Inc.

1-(3-Bromo-5-methoxy-phenyl)-ethylamine hydrochloride

  • MDL number MFCD31613996
  • Cas Number : 2270912-47-3
  • Molecular Weight : 266.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9413

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9413
Availability details for
Quantity
  • Availability
  • Add to Cart