COMH4211D1A6 Combi-Blocks, Inc.

2-Chloro-4-(2-methoxypyrimidin-5-yl)benzoic acid

  • MDL number MFCD31579448
  • Cas Number : 1819358-49-0
  • Molecular Weight : 264.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9416

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9416
Availability details for
Quantity
  • Availability
  • Add to Cart