COMH4211D1B0 Combi-Blocks, Inc.

Ethyl 4-methyl-2-(2-oxoimidazolidin-1-yl)thiazole-5-carboxylate

  • MDL number MFCD26387027
  • Cas Number : 1072806-73-5
  • Molecular Weight : 255.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9440

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9440
Availability details for
Quantity
  • Availability
  • Add to Cart