COMH4211D1B1 Combi-Blocks, Inc.

tert-Butyl 4-(2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenylsulfonyl)piperazine-1-carboxylate

  • MDL number MFCD31583299
  • Cas Number : 1628016-45-4
  • Molecular Weight : 482.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9441

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9441
Availability details for
Quantity
  • Availability
  • Add to Cart