COMH4211D1B4 Combi-Blocks, Inc.

[2-(3-Chloro-phenyl)-ethoxy]-acetic acid

  • MDL number MFCD19370532
  • Cas Number : 948894-91-5
  • Molecular Weight : 214.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9447

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9447
Availability details for
Quantity
  • Availability
  • Add to Cart