COMH4211D1BB Combi-Blocks, Inc.

Imidazole-1-carboxylic acid 3,4-dichloro-phenyl ester

  • MDL number MFCD31618982
  • Cas Number : 1205542-49-9
  • Molecular Weight : 257.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9459

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9459
Availability details for
Quantity
  • Availability
  • Add to Cart