COMH4211D1BD Combi-Blocks, Inc.

5-Bromo-n-cyclopropyl-2-(2,2,2-trifluoro-ethoxy)-benzamide

  • MDL number MFCD31628792
  • Cas Number : 2270910-58-0
  • Molecular Weight : 338.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9462

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9462
Availability details for
Quantity
  • Availability
  • Add to Cart