COMH4211D1C0 Combi-Blocks, Inc.

5-Bromo-2-iodo-benzoic acid tert-butyl ester

  • MDL number MFCD22919245
  • Cas Number : 450412-25-6
  • Molecular Weight : 383.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9466

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9466
Availability details for
Quantity
  • Availability
  • Add to Cart