COMH4211D1C3 Combi-Blocks, Inc.

5-(Benzyloxy)-2-methoxybenzoic acid

  • MDL number MFCD31618989
  • Cas Number : 84923-68-2
  • Molecular Weight : 258.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9472

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9472
Availability details for
Quantity
  • Availability
  • Add to Cart