COMH4211D1C5 Combi-Blocks, Inc.

[5-(3,4-Dichloro-phenyl)-thiophen-2-yl]-methanol

  • MDL number MFCD31619004
  • Cas Number : 2009598-15-4
  • Molecular Weight : 259.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9484

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9484
Availability details for
Quantity
  • Availability
  • Add to Cart