COMH4211D1CD Combi-Blocks, Inc.

4-Amino-n,n-diethyl-2-fluorobenzamide

  • MDL number MFCD16084599
  • Cas Number : 1188506-53-7
  • Molecular Weight : 210.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9498

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9498
Availability details for
Quantity
  • Availability
  • Add to Cart