COMH4211D1D3 Combi-Blocks, Inc.

1-[4-(Cyclopropylmethyl-carbamoyl)-phenyl]-5-propyl-1h-[1,2,3]triazole-4-carboxylic acid

  • MDL number MFCD31583040
  • Cas Number : 2270908-74-0
  • Molecular Weight : 328.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9508

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9508
Availability details for
Quantity
  • Availability
  • Add to Cart