COMH4211D1D5 Combi-Blocks, Inc.

1-(Morpholinomethyl)cyclopropanecarboxylic acid

  • MDL number MFCD19314028
  • Cas Number : 1257236-69-3
  • Molecular Weight : 185.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9514

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9514
Availability details for
Quantity
  • Availability
  • Add to Cart