COMH4211D1D7 Combi-Blocks, Inc.

(4-Hydroxy-pyrazol-1-yl)-phenyl-acetic acid methyl ester

  • MDL number MFCD31583307
  • Cas Number : 2103851-90-5
  • Molecular Weight : 232.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9519

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9519
Availability details for
Quantity
  • Availability
  • Add to Cart