COMH4211D1DA Combi-Blocks, Inc.

N-Cyclopropyl-2-[2-fluoro-4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-phenyl]-acetamide

  • MDL number MFCD31613951
  • Molecular Weight : 319.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9528

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9528
Availability details for
Quantity
  • Availability
  • Add to Cart