COMH4211D1DB Combi-Blocks, Inc.

4-Amino-3-(benzyloxy)-n,n-dimethylbenzamide

  • MDL number MFCD31583109
  • Cas Number : 1406094-94-7
  • Molecular Weight : 270.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9529

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9529
Availability details for
Quantity
  • Availability
  • Add to Cart