COMH4211D1DF Combi-Blocks, Inc.

5-Bromo-2-iodo-n-ethyl benzamide

  • MDL number MFCD19595465
  • Cas Number : 1295978-68-5
  • Molecular Weight : 354.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9542

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9542
Availability details for
Quantity
  • Availability
  • Add to Cart