COMH4211D1EA Combi-Blocks, Inc.

(4-Bromo-2-fluoro-5-methyl-phenyl)-pyrrolidin-1-yl-methanone

  • MDL number MFCD31579508
  • Cas Number : 2270907-54-3
  • Molecular Weight : 286.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9571

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9571
Availability details for
Quantity
  • Availability
  • Add to Cart