COMH4211D1EC Combi-Blocks, Inc.

2-Ethoxy-3-methylbenzaldehyde

  • MDL number MFCD23699165
  • Cas Number : 532965-62-1
  • Molecular Weight : 164.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9574

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9574
Availability details for
Quantity
  • Availability
  • Add to Cart