COMH4211D1ED Combi-Blocks, Inc.

4-(4-tert-Butoxycarbonylmethyl-piperazin-1-yl)-benzoic acid

  • MDL number MFCD31583321
  • Molecular Weight : 320.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9578

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9578
Availability details for
Quantity
  • Availability
  • Add to Cart