COMH4211D1F0 Combi-Blocks, Inc.

1-(1-Bromo-2,2,2-trifluoroethyl)-2,3,5-trichlorobenzene

  • MDL number MFCD31613795
  • Cas Number : 1416980-69-2
  • Molecular Weight : 342.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9581

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9581
Availability details for
Quantity
  • Availability
  • Add to Cart