COMH4211D1F3 Combi-Blocks, Inc.

3-(tert-Butyldimethylsilanyloxy)cyclohexanol

  • MDL number MFCD29054776
  • Cas Number : 1028748-71-1
  • Molecular Weight : 230.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9588

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9588
Availability details for
Quantity
  • Availability
  • Add to Cart