COMH4211D1F8 Combi-Blocks, Inc.

1-Ethanesulfonyl-1h-pyrazol-4-ol

  • MDL number MFCD31579525
  • Cas Number : 2270905-59-2
  • Molecular Weight : 176.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9595

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9595
Availability details for
Quantity
  • Availability
  • Add to Cart