COMH4211D1FF Combi-Blocks, Inc.

P-Tolyl 2,3,5-tri-o-benzoyl-1-thio-alpha-d-arabinofuranoside

  • MDL number MFCD31614083
  • Cas Number : 352438-38-1
  • Molecular Weight : 568.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9607

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9607
Availability details for
Quantity
  • Availability
  • Add to Cart