COMH4211D201 Combi-Blocks, Inc.

Trans-(4-hydroxymethyl-cyclohexyl)-carbamic acid allyl ester

  • MDL number MFCD30830140
  • Cas Number : 2167789-76-4
  • Molecular Weight : 213.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9609

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9609
Availability details for
Quantity
  • Availability
  • Add to Cart