COMH4211D205 Combi-Blocks, Inc.

(5-Hydroxy-2-methoxy-phenyl)-pyrrolidin-1-yl-methanone

  • MDL number MFCD31618901
  • Cas Number : 1052106-91-8
  • Molecular Weight : 221.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9619

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9619
Availability details for
Quantity
  • Availability
  • Add to Cart