COMH4211D214 Combi-Blocks, Inc.

N-[(3-Methylphenyl)methyl]piperidine-2-carboxamide

  • MDL number MFCD11849658
  • Cas Number : 189069-86-1
  • Molecular Weight : 232.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9648

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9648
Availability details for
Quantity
  • Availability
  • Add to Cart