COMH4211D217 Combi-Blocks, Inc.

Morpholino(4-vinylphenyl)methanone

  • MDL number MFCD31613788
  • Cas Number : 2681-27-8
  • Molecular Weight : 217.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9653

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9653
Availability details for
Quantity
  • Availability
  • Add to Cart